missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valproic Acid 1.0 mg/ml in Methanol, LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Valproic acid |
| CAS | 99-66-1 |
| Analyte or Component Names | Valproic acid |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C8 H16 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Valproic Acid, 2-Propylpentanoic acid, 2-Propylvaleric acid, 4-Heptanecarboxylic acid, 44089, Acetic acid, dipropyl-, DPA, Depakine, Dipropylacetic acid, Ergenyl, Mylproin, NSC 93819, Valproic acid, n-Dipropylacetic acid |
| InChI Formula | InChI=1S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10) |
| Solution Type | Single Solution |
| SMILES | CCCC(CCC)C(=O)O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction