missing translation for 'onlineSavingsMsg'
Learn More

Valinomycin, 90+%

Product Code. 15434719
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15434719 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15434719 Supplier Thermo Scientific Alfa Aesar Supplier No. J62312.MC
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2001-95-8 | C54H90N6O18 | 1111.34 g/mol

Valinomycin, is used as a potassium ionophore shown to induce cell death. Due to the ion-selective nature of valinomycin, it is used in ion-selective electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valinomycin, is used as a potassium ionophore shown to induce cell death. Due to the ion-selective nature of valinomycin, it is used in ion-selective electrodes.

Solubility
Soluble in DMSO at 10mg/ml. Insoluble in w
TRUSTED_SUSTAINABILITY

Specifications

Type Valinomycin
Physical Form Crystalline Powder
CAS 2001-95-8
Color White
Quantity 100 mg
Molecular Formula C54H90N6O18
MDL Number MFCD00005114
UN Number UN2811
Beilstein 78657
Merck Index 14,9910
Synonym NSC 122023
Solubility Information Soluble in DMSO at 10mg/ml; Insoluble in water
InChI Key FCFNRCROJUBPLU-UYBNATROSA-N
SMILES CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)C(NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C
IUPAC Name (3S,6S,12R,15S,18S,21R,24R,27S,30S,33R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nona(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
Molecular Weight (g/mol) 1111.34
PubChem CID 131668508
Formula Weight 1111.34
Percent Purity ≥90%
Chemical Name or Material Valinomycin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.