missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Valinomycin, 90%

Product Code. 10659384 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass Bottle
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10659384 10 mg Glass Bottle 10mg
10775213 50 mg Glass Bottle 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10659384 Supplier Thermo Scientific Chemicals Supplier No. 385120100
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2001-95-8
Molecular Formula C54H90N6O18
Molecular Weight (g/mol) 1111.34
MDL Number MFCD00005114
InChI Key FCFNRCROJUBPLU-UYBNATROSA-N
PubChem CID 131668508
IUPAC Name (3S,6S,12R,15S,18S,21R,24R,27S,30S,33R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nona(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
SMILES CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)C(NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C

Specifications

Type Valinomycin
Physical Form Crystalline Powder
Melting Point 190.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation +30° to +34° (20°C, 589nm) (c=1.6, benzene)
Color White
Quantity 10 mg
Assay Percent Range 90%
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: freely soluble in petroleum ether, ether, benzene,, chloroform, glacial acetic acid, butyl acetate and, acetone
Formula Weight 1111.35
Percent Purity 90%
Chemical Name or Material Valinomycin
Show More Show Less
compliance-icons
Product Identifier
  • Valinomycin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.