missing translation for 'onlineSavingsMsg'
Learn More

Melford Valinomycin

Product Code. 30018923
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018923 100 mg 100mg
30018922 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018923 Supplier Melford Supplier No. V120000.1
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Valinomycin is a naturally occurring cyclic dodecadepsipeptide produced by certain strains of Streptomyces bacteria.

Valinomycin is a naturally occurring cyclic dodecadepsipeptide produced by certain strains of Streptomyces bacteria. It is an ionophore, meaning it facilitates the transport of ions across biological membranes. Specifically, it acts as a potassium ionophore, transporting potassium ions across lipid bilayers. This activity disrupts ion gradients and membrane potentials in cells.

Widely used in research settings as a tool to study ion transport and membrane potential in cells. Its ability to selectively transport potassium ions makes it valuable in experiments exploring cellular electrophysiology. Valinomycin is used to manipulate ion concentrations and study mitochondrial function, as it can alter the electrochemical potential across the inner mitochondrial membrane.

Useful in a variety of experiments aimed at understanding cellular and mitochondrial physiology. It is often used alongside other tools to investigate ion transport mechanisms and study the effects of altered ion concentrations on cellular processes.

Specifications

Chemical Name or Material Valinomycin
CAS 2001-95-8
Quantity 100 mg
Molecular Formula C54H90N6O18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.