missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Validoxylamine A, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Validoxylamine A |
| CAS | 38665-10-0 |
| Molecular Formula | C14H25NO8 |
| Quantity | 1 mg |
| Synonym | 1,5,6-Trideoxy-5-(hydroxymethyl)-1-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)-2-cyclohexen-1-yl]amino]-D-chiro-inositol, (+)-Validoxylamine A |
| InChI Formula | InChI=1S/C14H25NO8/c16-3-5-1-7(11(20)13(22)9(5)18)15-8-2-6(4-17)10(19)14(23)12(8)21/h1,6-23H,2-4H2/t6-,7+,8+,9-,10-,11+,12+,13+,14+/m1/s1 |
| SMILES | OC[C@@H]1[C@@H](O)[C@H](O)[C@@H](O)[C@@H](N[C@@]2([H])C=C(CO)[C@@H](O)[C@H](O)[C@H]2O)C1 |
| IUPAC Name | (1S,2S,3R,6S)-4-(hydroxymethyl)-6-(((1S,2S,3S,4R,5R)-2,3,4-trihydroxy-5-(hydroxymethyl)cyclohexyl)amino)cyclohex-4-ene-1,2,3-triol |
| Molecular Weight (g/mol) | 335.35 |
| Formula Weight | 335.16 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction