missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valganciclovir-d5 Hydrochloride, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Valganciclovir-d5 Hydrochloride |
| Molecular Formula | C14H18D5ClN6O5 |
| Quantity | 1 mg |
| Synonym | 2-[(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]-3-hydroxypropyl-d5 Ester L-Valine Hydrochloride, RS 079070-194-d5, Ro 107-9070/194-d5, Valcyte-d5, (S)-Valganciclovir-d5 Hydrochloride |
| InChI Formula | InChI=1S/C14H22N6O5.ClH/c1-7(2)9(15)13(23)24-4-8(3-21)25-6-20-5-17-10-11(20)18-14(16)19-12(10)22;/h5,7-9,21H,3-4,6,15H2,1-2H3,(H3,16,18,19,22);1H/t8?,9-;/m0./s1/i3D2,4D2,8D; |
| SMILES | O=C1NC(N)=NC2=C1N=CN2COC(C([2H])([2H])O)([2H])C([2H])([2H])OC([C@@H](N)C(C)C)=O.Cl |
| Molecular Weight (g/mol) | 395.85 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction