missing translation for 'onlineSavingsMsg'
Learn More

Valeryl-L-carnitine Chloride, TRC

Product Code. 30454082
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454082 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454082 Supplier Toronto Research Chemicals Supplier No. TRCV09146525MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 162040-64-4
Molecular Formula C12 H23 N O4 . Cl
Quantity 25 mg
InChI Formula InChI=1S/C12H23NO4.ClH/c1-5-6-7-12(16)17-10(8-11(14)15)9-13(2,3)4;/h10H,5-9H2,1-4H3;1H/p-1/t10-;/m1./s1
SMILES [Cl-].CCCCC(=O)O[C@H](CC(=O)[O-])C[N+](C)(C)C
IUPAC Name (3R)-3-pentanoyloxy-4-(trimethylazaniumyl)butanoate;chloride
Molecular Weight (g/mol) 281.77626
Formula Weight 280.132
Chemical Name or Material Valeryl-L-carnitine Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.