missing translation for 'onlineSavingsMsg'
Learn More

Valeryl chloride, 98%

Product Code. 13408408 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13408408 1 kg Glass bottle 1kg
10194320 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13408408 Supplier Thermo Scientific Acros Supplier No. 169120010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 638-29-9 | C5H9ClO | 120.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-29-9
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00000757
InChI Key XGISHOFUAFNYQF-UHFFFAOYSA-N
Synonym valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride
PubChem CID 61186
IUPAC Name pentanoyl chloride
SMILES CCCCC(Cl)=O

Specifications

Melting Point -110.0°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 125.0°C to 127.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3COCl
Refractive Index 1.4190 to 1.4210
Quantity 1 kg
Beilstein 02,301
Specific Gravity 0.99
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in most common organic solvents
Formula Weight 120.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Valeryl chloride, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Valeryl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.