missing translation for 'onlineSavingsMsg'
Learn More

Valeryl chloride, 98%

Product Code. 11445247
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445247 50 g 50g
11455247 250 g 250g
11435247 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445247 Supplier Thermo Scientific Alfa Aesar Supplier No. B22637.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 638-29-9 | C5H9ClO | 120.58 g/mol

Valeryl chloride is an intermediate for pharmaceuticals, agrochemicals, surfactants, fragrances, electronic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valeryl chloride is an intermediate for pharmaceuticals, agrochemicals, surfactants, fragrances, electronic chemicals.

Solubility
Hydrolyzes in water. Soluble in ether, chloroform.

Notes
Moisture sensitive. Stay away from oxidizing agents, bases, alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-29-9
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00000757
InChI Key XGISHOFUAFNYQF-UHFFFAOYSA-N
Synonym valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride
PubChem CID 61186
IUPAC Name pentanoyl chloride
SMILES CCCCC(Cl)=O

Specifications

Melting Point -110°C
Boiling Point 125°C to 127°C
Flash Point 32°C (89°F)
Odor Acrid
Quantity 50 g
UN Number UN2502
Beilstein 506293
Formula Weight 120.58
Percent Purity 98%
Chemical Name or Material Valeryl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.