missing translation for 'onlineSavingsMsg'
Learn More

Valerophenone, 99%

Product Code. 11478793
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478793 100 g 100g
11468793 25 g 25g
11488793 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478793 Supplier Thermo Scientific Alfa Aesar Supplier No. A10525.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1009-14-9 | C11H14O | 162.23 g/mol

Valerophenone is used as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valerophenone is used as intermediates of liquid crystals.

Solubility
Insoluble in water.

Notes
 Incompatible with strong oxidizing agents, acids, bases, plastics. Store at room temperature with proper ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1009-14-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00009480
InChI Key XKGLSKVNOSHTAD-UHFFFAOYSA-N
Synonym valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci
PubChem CID 66093
ChEBI CHEBI:36812
IUPAC Name 1-phenylpentan-1-one
SMILES CCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point -9°C
Density 0.978
Boiling Point 244°C to 245°C
Flash Point 102°C (215°F)
Assay Percent Range 99%
Refractive Index 1.5143
Quantity 100 g
Beilstein 1907717
Solubility Information Insoluble in water.
Formula Weight 162.23
Percent Purity 99%
Chemical Name or Material Valerophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.