missing translation for 'onlineSavingsMsg'
Learn More

Valeronitrile, 99%

Product Code. 11474053
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474053 500 mL 500mL
11473890 25 mL 25mL
11464053 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474053 Supplier Thermo Scientific Alfa Aesar Supplier No. A10306.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-59-8 | C5H9N | 83.134 g/mol

Valeronitrile is used in the preparation of valeric acid. It is also used to enhance the nitrilase activity in many strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valeronitrile is used in the preparation of valeric acid. It is also used to enhance the nitrilase activity in many strains.

Solubility
Soluble in water at 20°C 10g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents, acids, bases and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-59-8
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00001974
InChI Key RFFFKMOABOFIDF-UHFFFAOYSA-N
Synonym valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile
PubChem CID 8061
IUPAC Name pentanenitrile
SMILES CCCCC#N

Specifications

Melting Point -96°C
Density 0.797
Boiling Point 139°C to 141°C
Flash Point 38°C (100°F)
Odor Odorless
Refractive Index 1.397
Quantity 500 mL
UN Number UN1992
Beilstein 1736706
Merck Index 14,9905
Solubility Information Soluble in water at 20°C 10g/L. Soluble in alcohol.
Formula Weight 83.13
Percent Purity 99%
Chemical Name or Material Valeronitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.