missing translation for 'onlineSavingsMsg'
Learn More

Valeronitrile, 98%

Product Code. 10403965 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10403965 500 mL Glass bottle 500mL
10022090 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10403965 Supplier Thermo Scientific Acros Supplier No. 167945000
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-59-8 | C5H9N | 83.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-59-8
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00001974
InChI Key RFFFKMOABOFIDF-UHFFFAOYSA-N
Synonym valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile
PubChem CID 8061
IUPAC Name pentanenitrile
SMILES CCCCC#N

Specifications

Melting Point -96.0°C
Color Colorless
Density 0.7900g/mL
Boiling Point 139.0°C to 141.0°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3CN
Refractive Index 1.3960 to 1.3980
Quantity 500 mL
Beilstein 02, 301
Merck Index 15, 10091
Specific Gravity 0.79
Formula Weight 83.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Valeronitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Valeronitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.