Learn More
Valeronitrile, 98%, ACROS Organics™
Brand: Acros Organics 167941000
Code : 29
Additional Details : CAS Number : 110-59-8 Weight : 0.12900kg Transport : UN number : 1992 Chem class : 3 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 100mL |
Chemical Identifiers
110-59-8 | |
83.13 | |
RFFFKMOABOFIDF-UHFFFAOYSA-N | |
8061 | |
CCCCC#N |
C5H9N | |
MFCD00001974 | |
valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile | |
pentanenitrile |
Specifications
Valeronitrile | |
40°C | |
Glass bottle | |
0.79 | |
139.0°C to 141.0°C | |
-96.0°C | |
110-59-8 | |
C5H9N | |
MFCD00001974 | |
15, 10091 | |
RFFFKMOABOFIDF-UHFFFAOYSA-N | |
pentanenitrile | |
8061 | |
Liquid |
0.7900g/mL | |
Authentic | |
1.3960 to 1.3980 | |
0.15% max. (K.F.) | |
Colorless | |
100mL | |
97.5% min. (GC) | |
CH3(CH2)3CN | |
02, 301 | |
valeronitrile, butyl cyanide, n-valeronitrile, 1-cyanobutane, 1-butyl cyanide, n-butyl cyanide, pentanonitrile, unii-x44h3r47d4, n-butylcyanide, n-pentanenitrile | |
CCCCC#N | |
83.13 | |
83.13 | |
98% |
Safety and Handling
- Valeronitrile
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /