missing translation for 'onlineSavingsMsg'
Learn More

delta-Valerolactone, 99%

Product Code. 10275780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10275780 250 g Glass bottle 250g
10666752 50 g Glass bottle 50g
10647364 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10275780 Supplier Thermo Scientific Acros Supplier No. 162712500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 542-28-9 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-28-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00006645
InChI Key OZJPLYNZGCXSJM-UHFFFAOYSA-N
Synonym delta-valerolactone, tetrahydro-2h-pyran-2-one, 5-valerolactone, 2h-pyran-2-one, tetrahydro, tetrahydropyran-2-one, tetrahydro-2-pyranone, penta-1,5-lactone, .delta.-valerolactone, .delta.-valeryllactone, tetrahydro-pyran-2-one
PubChem CID 10953
ChEBI CHEBI:16545
IUPAC Name oxan-2-one
SMILES O=C1CCCCO1

Specifications

Melting Point -13.0°C
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 230.0°C
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4560 to 1.4590
Quantity 250 g
Beilstein 17, 235
Specific Gravity 1.1
Solubility Information Solubility in water: miscible
Formula Weight 100.12
Percent Purity 99%
Concentration or Composition (by Analyte or Components) May contain variable amounts of polymerized product
Physical Form Liquid
Chemical Name or Material δ-Valerolactone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • delta-Valerolactone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.