missing translation for 'onlineSavingsMsg'
Learn More

Valeric Acid, TRC

Product Code. 30456971
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456971 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456971 Supplier Toronto Research Chemicals Supplier No. TRCV0914151ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Valeric Acid
CAS 109-52-4
Molecular Formula C5 H10 O2
Quantity 1 mL
Synonym Valeric acid, Pentanoic acid, Valeric acid (8CI), 1-Butanecarboxylic acid, NSC 406833, Propylacetic acid, n-Pentanoic acid, n-Valeric acid
InChI Formula InChI=1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)
SMILES CCCCC(=O)O
IUPAC Name pentanoic acid
Molecular Weight (g/mol) 102.13
Formula Weight 102.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.