missing translation for 'onlineSavingsMsg'
Learn More

Valeramide, 97%, Thermo Scientific Chemicals

Catalog Number p-211650
Click to view available options
Quantity:
2.5 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
626-97-1
C5H11NO
101.15
MFCD00041895
IPWFJLQDVFKJDU-UHFFFAOYSA-N
valeramide, n-valeramide, pentanimidic acid, unii-tkl9tn43qk, tkl9tn43qk, ksc353i5p, ipwfjlqdvfkjdu-uhfffaoysa, valeramide, pharmaceutical impurity standard
12298
CHEBI:16459
pentanamide
CCCCC(=O)N
This item is not returnable. View return policy
626-97-1
C5H11NO
101.15
MFCD00041895
IPWFJLQDVFKJDU-UHFFFAOYSA-N
valeramide, n-valeramide, pentanimidic acid, unii-tkl9tn43qk, tkl9tn43qk, ksc353i5p, ipwfjlqdvfkjdu-uhfffaoysa, valeramide, pharmaceutical impurity standard
12298
CHEBI:16459
pentanamide
CCCCC(=O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 101°C to 106°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3CONH2
Quantity 2.5 g
Formula Weight 101.15
Percent Purity 97%
Physical Form Shiny Powder or Flakes
Chemical Name or Material Valeramide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Valeramide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'