missing translation for 'onlineSavingsMsg'
Learn More

Valeraldehyde (Pentanal), TRC

Product Code. 30136535
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136535 1 g 1g
30005872 10 g 10g
30009746 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136535 Supplier Toronto Research Chemicals Supplier No. TRCV0912951G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-62-3
Molecular Formula C5 H10 O
Molecular Weight (g/mol) 86.13
Synonym Valeraldehyde, Pentanal, Valeraldehyde (8CI), NSC 35404, Pentylaldehyde, Valeral, Valeric acid aldehyde, Valeric aldehyde, n-Pentanal, n-Valeraldehyde
IUPAC Name pentanal
SMILES CCCCC=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H10O/c1-2-3-4-5-6/h5H,2-4H2,1H3
Formula Weight 86.0732
Chemical Name or Material n-Valeraldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.