missing translation for 'onlineSavingsMsg'
Learn More

Valeraldehyde, 97%

Product Code. 10356550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10356550 250 mL Glass bottle 250mL
10715041 100 mL Glass bottle 100mL
10666164 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10356550 Supplier Thermo Scientific Acros Supplier No. 149562500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-62-3 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-62-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00007026
InChI Key HGBOYTHUEUWSSQ-UHFFFAOYSA-N
Synonym valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde
PubChem CID 8063
ChEBI CHEBI:84069
IUPAC Name pentanal
SMILES CCCCC=O

Specifications

Melting Point -92.0°C
Density 0.8070g/mL
Boiling Point 103.0°C
Flash Point 12°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3CHO
Refractive Index 1.3900 to 1.3950
Quantity 250 mL
Beilstein 01, 676
Merck Index 15, 10088
Specific Gravity 0.807
Solubility Information Solubility in water: 14g/L (20°C). Other solubilities: soluble in alcohol, ether, propylene glycol and, many organic solvents
Viscosity 0.6 mPa.s (20°C)
Formula Weight 86.13
Percent Purity 97%
Chemical Name or Material Valeraldehyde, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.