Learn More
Emma Safety Footwear Valentino (Frontier 164) Safety Shoes
Brazilian 2-tone full grain leather combined with nubuck adds to the stylish look.
Brand: Emma Safety Footwear MM16409046
Additional Details : Weight : 2.29900kg
Description
- S3-Safety shoe with sole plate for wet work
- TPU Outsole
- Full grain leather
- ESD Certified
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Hydro-Tec™ Sanitized Silver & Wellness leather | |
46 |
Laces | |
Valentino (Frontier 164) | |
Leather, Nubuck | |
Steel |