Learn More
Thermo Scientific™ Valco™ Injector Model C6W
Use the Thermo Scientific™ Valco™ Injector Model C6W as an injector or switching valve.
£29.46 - £37.01
Specifications
No. of Ports 6 Ports
View More Specs
Products 4
Product Code Brand Description Volume (Metric) Sample Price Quantity & Availability  
Product Code Brand Description Volume (Metric) Sample Price Quantity & Availability  
11563980
View Documents Promotion Details
Thermo Scientific™
SL100CW
Sample injector loop, stainless steel 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11573980
View Documents Promotion Details
Thermo Scientific™
SL10CW
Sample injector loop, stainless steel 10 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11503990
View Documents Promotion Details
Thermo Scientific™
SL50CW
Sample injector loop, stainless steel 50 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11513990
View Documents Promotion Details
Thermo Scientific™
SL5CW
Sample injector loop, stainless steel 5 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Use the Thermo Scientific™ Valco™ Injector Model C6W as an injector or switching valve. Available with manual or a microelectric actuator. Has 1/16 in. fittings and six 0.010 inch ports.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
6 Ports
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.