missing translation for 'onlineSavingsMsg'
Learn More

Valbenazine, MedChemExpress

Product Code. 16493207
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493207 50 mg 50mg
16477176 100 mg 100mg
16469717 5 mg 5mg
16467188 10 mg 10mg
16443546 25 mg 25mg
16430286 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16493207 Supplier Medchem Express Supplier No. HY16771
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Valbenazine (NBI-98854) is a vesicular monoamine transporter 2 (VMAT2) inhibitor with the Ki of 110-190 nM.

Valbenazine (NBI-98854) is a vesicular monoamine transporter 2 (VMAT2) inhibitor with the Ki of 110-190 nM.

Specifications

Chemical Name or Material Valbenazine
CAS 1025504-45-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H38N2O4
Quantity 50 mg
Synonym NBI-98854
Solubility Information DMSO : 50 mg/mL (119.45 mM; Need ultrasonic)
SMILES O=C([C@@H](N)C(C)C)O[C@@H]1C[C@]2([H])C3=CC(OC)=C(OC)C=C3CCN2C[C@H]1CC(C)C
Molecular Weight (g/mol) 418.57
Formula Weight 418.57
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.