missing translation for 'onlineSavingsMsg'
Learn More

Valacyclovir hydrochloride, MedChemExpress

Product Code. 16437338
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16437338 10 mg 10mg
16413357 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16437338 Supplier Medchem Express Supplier No. HY17425A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Valacyclovir hydrochloride (Valaciclovir hydrochloride) is an orally active antiviral drug for herpes simplex, herpes zoster, and herpes B. Valacyclovir hydrochloride inhibits HSV-1 W (50=2.9 μg/ml). Valacyclovir hydrochloride is a prodrug of Aciclovir (HY-17422) .

Valacyclovir hydrochloride (Valaciclovir hydrochloride) is an orally active antiviral drug for herpes simplex, herpes zoster, and herpes B. Valacyclovir hydrochloride inhibits HSV-1 W (50=2.9 μg/ml). Valacyclovir hydrochloride is a prodrug of Aciclovir (HY-17422) .

Specifications

Chemical Name or Material Valacyclovir hydrochloride
Physical Form Solid
CAS 124832-27-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C13H21ClN6O4
For Use With (Application) COVID-19-anti-virus
Synonym Valaciclovir hydrochloride
Solubility Information H2O : 100 mg/mL (277.16 mM; Need ultrasonic) ∣DMSO : 43.33 mg/mL (120.09 mM; Need ultrasonic)
SMILES N[C@@H](C(C)C)C(OCCOCN1C=NC2=C1N=C(N)NC2=O)=O.Cl
Molecular Weight (g/mol) 360.8
Formula Weight 360.8
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.