missing translation for 'onlineSavingsMsg'
Learn More

Val-Cit-PAB-MMAE, MedChemExpress

Product Code. 16435717
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16435717

Brand: Medchem Express HY100374

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Val-Cit-PAB-MMAE is a drug-linker conjugate for ADC. Val-Cit-PAB-MMAE contains the ADCs linker (peptide Val-Cit-PAB) and a potent tubulin inhibitor MMAE (HY-15162). MMAE a potent mitotic inhibitor by inhibiting tubulin polymerization.

Val-Cit-PAB-MMAE is a drug-linker conjugate for ADC. Val-Cit-PAB-MMAE contains the ADCs linker (peptide Val-Cit-PAB) and a potent tubulin inhibitor MMAE (HY-15162). MMAE a potent mitotic inhibitor by inhibiting tubulin polymerization.

Specifications

Chemical Name or Material Val-Cit-PAB-MMAE
Physical Form Solid
CAS 644981-35-1
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C58H94N10O12
Solubility Information DMSO : 50 mg/mL (44.51 mM; Need ultrasonic)
SMILES CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C)[C@@H](O)C2=CC=CC=C2)=O)OC)=O)OC)N(C([C@H](C(C)C)NC([C@H](C(C)C)N(C(OCC3=CC=C(NC([C@H](CCCNC(N)=O)NC([C@H](C(C)C)N)=O)=O)C=C3)=O)C)=O)=O)C
Molecular Weight (g/mol) 1123.43
Formula Weight 1123.43
Percent Purity 95.73%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.