missing translation for 'onlineSavingsMsg'
Learn More

VAL-083, MedChemExpress

Product Code. 16487068
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487068

Brand: Medchem Express HY16513

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VAL-083 is an alkylating agent that creates N7 methylation on DNA, with antitumor activity.

VAL-083 is an alkylating agent that creates N7 methylation on DNA, with antitumor activity.

Specifications

Chemical Name or Material VAL-083
Physical Form Solid
CAS 23261-20-3
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C6H10O4
For Use With (Application) Cancer-programmed cell death
Synonym Dianhydrodulcitol Dianhydrogalactitol
Solubility Information DMF : ≥ 100 mg/mL (684.28 mM) ∣H2O : 50 mg/mL (342.14 mM; Need ultrasonic)
SMILES O[C@H]([C@H](O)[C@]1(OC1)[H])[C@@]2([H])CO2
Molecular Weight (g/mol) 146.14
Formula Weight 146.14
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.