missing translation for 'onlineSavingsMsg'
Learn More

Vadadustat, MedChemExpress

Product Code. 16483318
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483318 10 mg 10mg
16464566 100 mg 100mg
16449856 50 mg 50mg
16406047 5 mg 5mg
16400056 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16483318 Supplier Medchem Express Supplier No. HY101277
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Vadadustat (PG-1016548) is a titratable, oral hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor. Vadadustat is an erythropoiesis-stimulating agent and has the potential for anemia treatment in chronic kidney disease in vivo.

Vadadustat (PG-1016548) is a titratable, oral hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor. Vadadustat is an erythropoiesis-stimulating agent and has the potential for anemia treatment in chronic kidney disease in vivo.

Specifications

Chemical Name or Material Vadadustat
CAS 1000025-07-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H11ClN2O4
Quantity 10 mg
Synonym PG-1016548 AKB-6548
Solubility Information DMSO : 100 mg/mL (326.05 mM; Need ultrasonic)
SMILES O=C(O)CNC(C1=NC=C(C2=CC=CC(Cl)=C2)C=C1O)=O
Molecular Weight (g/mol) 306.7
Formula Weight 306.7
Percent Purity 99.25%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.