missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Thermo Scientific VACUUM OVEN 128L 200°C SOLVENT

Brand:  Thermo Scientific 51014547

Thermo Scientific VACUUM OVEN 128L 200°C SOLVENT

Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Product Code. 10312622

  • £19495.00 / Each
Only null left
Estimated Shipment: 09-02-2026
Add to basket
This item is not returnable. View return policy

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.