Learn More
Day-Impex Vacuum-insulated Stainless Steel Dewar Flasks
Product Code. 11773619
Product Code. 11773619 Supplier Day-Impex Supplier No. 320002
  • ONLINE EXCLUSIVE PRICE
    £455.94
    £364.44 / Pack of 1
    Save £91.50
    20% Off
 Restricted Item
Estimated Shipment: 18-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Dilvac™ vacuum-insulated flasks are entirely stainless steel. Lightweight and compact in design they are suitable for ultra-low or very high temperature applications. Available in a range of capacities from 0.5 L to 6 L.
  • Vacuum-insulated
  • Lightweight stainless steel
  • Ideal for storage or transportation
  • Suitable for ultra-low and very high temperatures