Learn More
Day-Impex Vacuum-insulated Stainless Steel Dewar Flasks
Product Code. 11793619
Product Code. 11793619 Supplier Day-Impex Supplier No. 320004
  • ONLINE EXCLUSIVE PRICE
    £715.41
    £416.50 / Pack of 1
    Save £298.91
    41% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Dilvac™ vacuum-insulated flasks are entirely stainless steel. Lightweight and compact in design they are suitable for ultra-low or very high temperature applications. Available in a range of capacities from 0.5 L to 6 L.
  • Vacuum-insulated
  • Lightweight stainless steel
  • Ideal for storage or transportation
  • Suitable for ultra-low and very high temperatures