Learn More
BUCHI Vacuum Controller V-855
Product Code. 10217462 Shop All Buchi Products
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Product Code. 10217462
Brand: BUCHI 047232
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use this vacuum controller with rotary evaporation systems. BUCHI™ Vacuum Controller V-855 features automatic distillation functions based on either vapor pressure or vapor temperature (latter requires optional autodistillation probe).
V-801 EasyVac
EasyVac system is a dedicated module designed to work specifically with the V-700 or V-710 pumps to allow automatic distillations with any rotary evaporator. The module gradually lowers the vacuum until solvent vapour is detected. Once all vapour has been evaporated, the controller then looks for other solvents. The process is repeated until all solvents have been distilled off.
V-850 controller
Semi-automatic controller. With solvent library of 43 common solvents and timer function. Full manual control also possible, including vacuum release.
V-855 controller
Automatic controller. As V-850 plus automatic distillation functions based on either vapour pressure or vapour temperature (latter requires optional autodistillation probe).
  • Vacuum controllers are multi-function units with a bright LCD digital and analogue graphics display
  • Range of pre-loaded applications
  • All units fully compatible with Büchi and other brands of vacuum pumps
  • Retort stand mounting
Specifications
Type Vacuum Controller V-855
Quantity 1 Each
Electrical Requirements 85/264 V 50/60 Hz
Voltage 85/264 V
Frequency 50/60 Hz