missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Vacuum Connecting Hose 3M W/Connection Accessories (Not Required With Pump Module), For Vo49
Shop All Memmert ProductsDescription
- Vacuum connecting hose from oven to Memmert pump, 3m
- Includes optimized connection accessories
- Partially stainless steel
Specifications
Specifications
| For Use With (Equipment) | Cooled vacuum drying oven Vocool |
| Type | Vacuum Connecting Hose |
| Material | Stainless Steel |
| Length (Metric) | 300 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?