missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Vacutainer™ Syringe Critical Care Blood Collection
Brand: BD 364327
Code : 02
Additional Details : Weight : 3.50000kg
Description
With needle BD preset, luer-lok with bd Hemogard™ tip cap 23G needle volume 3mL BD VacutainerSpecifications
23 G | |
BD Hemogard™ type tip cap | |
Luer Lock |
Critical care testing on whole blood | |
3 mL |