missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ SST™ II Advance Tubes

Product Code. 12917696
Change view
Click to view available options
Capacity (Metric):
3.5 mL
5 mL
8.5 mL
Closure Color:
Gold
Red
Label Type:
Block
Paper
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Label Type unitSize
12917696 5 mL Gold Block Pack of 1000
12987686 3.5 mL Gold Block Pack of 1000
12927696 8.5 mL Gold Block Pack of 1000
12997686 3.5 mL Red Block Pack of 1000
12907696 5 mL Red Paper Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12917696 Supplier BD Supplier No. 367954
  • ONLINE EXCLUSIVE PRICE
    £863.45
    £677.53 / Pack of 1000
    Save £185.92
    21% Off
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Prevents the contamination of the serum from the separated cellular components. BD Vacutainer™ SST™ II Advance Tubes feature an inert, stable gel that separates the serum and the blood clot during centrifugation. The type of gel used enables short centrifugation times.

Using BD SST™ II Advance, routine analytes in clinical chemistry, such as potassium and glucose, are still stable after a week of storage at 2–8°C. Clinical evaluation of special chemistries (such as therapeutic drugs, proteins, peptides, steroids and vitamins) demonstrates a high degree of stability with the acrylic gel in these tubes, which is a distinct advantage during transport and storage.

Advantages of gel tubes versus non-gel tubes:

  • Stable barrier between serum and clotted blood, therefore better analyte stability and sample quality
  • Optimization of the work flow: short centrifugation time, sample processing and archiving in the primary tube
  • No possibility of misidentification due to the use of secondary tubes

Tube usage guidelines:

  • Clotting Times: the minimum recommended coagulation time for patients who have not received anti-coagulation treatment is 30 minutes. Tubes contain a clot activator of silica particles.
  • Centrifugation Conditions: 1300–2000 g for 10 minutes or alternately 3000 g for 5 minutes, at 18–25°C.
  • Effects of Temperature: cooling of the tube during centrifuging can affect the movement capability of the gel. Optimum separation of serum and coagulated blood is achieved at 20–25°C.
  • Store at 4–25°C and protected from direct sunlight during storage.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Serum Separator Tube
Additive Silica
Material PET
Sterility Sterile
Closure Type Hemogard
Capacity (Metric) 5 mL
Closure Color Gold
Height (Metric) 100 mm
For Use With (Application) Used to obtain and separate a serum sample
Label Type Block
Diameter (Metric) Outer 13 mm
Certifications/Compliance CE
Show More Show Less
compliance-icons
Product Identifier
  • TUBE SST II PLH 13X100 5.0 BLBL CE GLD
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 2
Hazard Statement
  • H350-May cause cancer.
  • H361d-Suspected of damaging the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methyl benzene, quartz, 2,2,4,4,6,6,8,8-octamethyl-cyclotetrasiloxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.