missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Plastic Urinalysis Tubes

Product Code. 12977766
Click to view available options
Volume (Metric):
4 mL
6 mL
8 mL
9.5 mL
10 mL
11 mL
Bottom Shape:
Conical
Round
Closure Type:
Conventional Rubber Stopper
Hemogard™
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 12977766

Brand: BD 364938

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For the collection and transport of urine specimens. BD Vacutainer™ Plastic Urinalysis Tubes are sterile evacuated tubes with or without preservative to be used with BD collection devices. Available in many sizes with a round or conical bottom.

BD Vacutainer™ urine products for the collection and transport of urine specimens offer the same level of safety and standardization as BD blood collection products.
  • Designed to provide accurate patient results with reduced risk of healthcare worker exposure to hazardous specimens
  • Used with BD collection devices, specimen cups, 24 hour 3L containers and transfer straws
  • Leak-proof urine tubes can safely transport urine specimens to the laboratory for analysis
  • Options include bottom shape (round or conical) and cap style (conventional or Hemogard™)
  • Most sizes are preservative/additive free
  • 8mL tube (mfr. no. 364938) contains a preservative: 0.4% Chlorhexidine, 5.6% Ethyl Paraben and 94% Sodium Propionate
TRUSTED_SUSTAINABILITY

Specifications

Product Type Urinalysis Tube
Closure Color Yellow
Closure Type Conventional Rubber Stopper
Disposable Single-use
Material PET
Sterility Sterile
Additive No additive
Bottom Shape Round
For Use With (Application) Urinalysis
For Use With (Equipment) Urine collection devices (cups, transfer straws, 24 hr. containers)
Label Description Paper
No. per Pack 100 Pk.
Storage Requirements Normal Conditions
Volume (Metric) 10 mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.