missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Plastic Urinalysis Tubes

Product Code. 12947766
Click to view available options
Volume (Metric):
4 mL
6 mL
8 mL
9.5 mL
10 mL
11 mL
Bottom Shape:
Conical
Round
Closure Type:
Conventional Rubber Stopper
Hemogard™
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 12947766

Brand: BD 368501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For the collection and transport of urine specimens. BD Vacutainer™ Plastic Urinalysis Tubes are sterile evacuated tubes with or without preservative to be used with BD collection devices. Available in many sizes with a round or conical bottom.

BD Vacutainer™ urine products for the collection and transport of urine specimens offer the same level of safety and standardization as BD blood collection products.
  • Designed to provide accurate patient results with reduced risk of healthcare worker exposure to hazardous specimens
  • Used with BD collection devices, specimen cups, 24 hour 3L containers and transfer straws
  • Leak-proof urine tubes can safely transport urine specimens to the laboratory for analysis
  • Options include bottom shape (round or conical) and cap style (conventional or Hemogard™)
  • Most sizes are preservative/additive free
  • 8mL tube (mfr. no. 364938) contains a preservative: 0.4% Chlorhexidine, 5.6% Ethyl Paraben and 94% Sodium Propionate
TRUSTED_SUSTAINABILITY

Specifica

Product Type Urinalysis Tube
Closure Color Beige
Closure Type Hemogard™
Disposable Single-use
Material PET
Sterility Sterile
Additive No additive
Bottom Shape Round
For Use With (Application) Urinalysis
For Use With (Equipment) Urine collection devices (cups, transfer straws, 24 hr. containers)
Label Description Paper
Volume (Metric) 6 mL
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato