missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Hemogard™ Closure Plastic K2-Edta Tube

Product Code. 10371709
Change view
Click to view available options
Capacity (Metric):
6 mL
10 mL
Unit Size:
Pack of 100
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10371709 10 mL Pack of 100
12957666 6 mL Pack of 1000
10331254 10 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10371709 Supplier BD Supplier No. 367525
  • ONLINE EXCLUSIVE PRICE
    £33.57
    £28.83 / Pack of 100
    Save £4.74
    14% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to obtain a whole blood or EDTA plasma sample. Vacutainer™ Hemogard™ Closure Plastic K2-Edta Tube contains K2EDTA which is a liquid solution that is spray coated onto the interior surface of the BD Vacutainer Plus Tubes.

Lavender Hemogard™ closure
Paper label
TRUSTED_SUSTAINABILITY

Specifications

Product Type K2 EDTA Tube
Additive K2 EDTA
Material Plastic
Closure Type Hemogard
Capacity (Metric) 10 mL
Closure Color Lavender
Height (Metric) 100 mm
For Use With (Application) Used to obtain a whole blood or EDTA plasma sample
Label Type Paper
Diameter (Metric) Outer 16 mm
Certifications/Compliance ISO 6710
Show More Show Less
compliance-icons
Product Identifier
  • TUBE EDTA PLH 16X100 10.0 PLBLCE LAV
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.