Learn More
BD Vacutainer™ Hemogard Closure Plastic K2-Edta Tube
Used to obtain a whole blood or EDTA plasma sample.   Vacutainer™ Hemogard Closure Plastic K2-Edta Tube contains K2EDTA which is a liquid solution that is spray coated onto the interior surface of the BD Vacutainer Plus Tubes.
Brand:  BD VS367873
Product Code. 12834801
  • £70.42 / Pack of 100
Description
Lavender Hemogard closure
Paper label