missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Vacutainer™ Eclipse™ PST™ II Plasma Separator Tubes
Click to view available options
Capacity (Metric):
3 mL
4.5 mL
8 mL
Tube Size:
13 x 75 mm
13 x 100 mm
16 x 100 mm
Unit Size:
Pack of 1000
Description
During centrifugation of Eclipse PST II tubes, an inert gel separates serum and red blood cells, preventing the contamination of plasma from the separated cellular components. Routine analytes (e.g., potassium, glucose) and special chemistries (e.g., therapeutic drugs, proteins, peptides, steroids and vitamins) demonstrate a high degree of stability.
- The main advantages of gel tubes versus non-gel tubes are:
- Stable barrier between plasma and red blood cells
- Better analyte stability and sample quality
- Optimized workflow: short centrifugation time, sample processing and archiving in one tube
- No possibility of confusion due to the use of secondary tubes
- Storage: Store at 4-25°C, protected from direct sunlight; optimum separation is achieved at 20-25°C
- Mixing the tube: Invert tube 8-10 times immediately after collecting sample to avoid microclotting
- Centrifugation conditions at 18-25°C: 1300-2000g for 10 minutes or 3000g for 5 minutes
Specifications
Specifications
| Product Type | PST II Tube |
| Additive | Lithium Heparin and Plasma Separator |
| Material | PET |
| Sterility | Sterile |
| Closure Type | Hemogard |
| Capacity (Metric) | 3 mL |
| For Use With (Equipment) | Vacutainer™ Needle and Tube Holder; Centrifugator |
| Closure Color | Light Green |
| Tube Size | 13 x 75 mm |
| For Use With (Application) | Used to obtain and separate a serum sample |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction