missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Eclipse™ PST™ II Plasma Separator Tubes

Product Code. 12917686
Click to view available options
Capacity (Metric):
3 mL
4.5 mL
8 mL
Tube Size:
13 x 75 mm
13 x 100 mm
16 x 100 mm
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 12917686

Brand: BD 367377

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Features an inert separation gel and a spray-dried Lithium Heparin anticoagulant for clinical chemistry analyses. BD Vacutainer™ Eclipse™ PST™ II Plasma Separator Tubes are sterile, evacuated tubes that enable separation, storage and transport of plasma samples without the use of additional tubes.

During centrifugation of Eclipse PST II tubes, an inert gel separates serum and red blood cells, preventing the contamination of plasma from the separated cellular components. Routine analytes (e.g., potassium, glucose) and special chemistries (e.g., therapeutic drugs, proteins, peptides, steroids and vitamins) demonstrate a high degree of stability.

  • The main advantages of gel tubes versus non-gel tubes are:
    • Stable barrier between plasma and red blood cells
    • Better analyte stability and sample quality
    • Optimized workflow: short centrifugation time, sample processing and archiving in one tube
    • No possibility of confusion due to the use of secondary tubes
  • Storage: Store at 4-25°C, protected from direct sunlight; optimum separation is achieved at 20-25°C
  • Mixing the tube: Invert tube 8-10 times immediately after collecting sample to avoid microclotting
  • Centrifugation conditions at 18-25°C: 1300-2000g for 10 minutes or 3000g for 5 minutes
TRUSTED_SUSTAINABILITY

Specifications

Product Type PST II Tube
Additive Lithium Heparin and Plasma Separator
Material PET
Sterility Sterile
Closure Type Hemogard
Capacity (Metric) 8 mL
For Use With (Equipment) Vacutainer™ Needle and Tube Holder; Centrifugator
Closure Color Light Green
Tube Size 16 x 100 mm
For Use With (Application) Used to obtain and separate a serum sample
Label Type Block
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.