missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Vacutainer™ Crossmatch Serum Tubes
Click to view available options
Capacity (Metric):
4 mL
6 mL
Additive:
K2 EDTA (Spray)
Silica (Clot Activator)
Tube Size:
13 x 75 mm
13 x 100 mm
Unit Size:
Pack of 1000
Description
The BD Vacutainer™ system is a closed system optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process. The BD Vacutainer™ Crossmatch tube is identified by:
- Sterile evacuated tube made of durable PET plastic
- Color coded pink Hemogard™ Cap designed for use with Vacutainer needles and tube holders
- Specialized label is larger than a typical block label to allow more information to be recorded
- Mixing: Tubes should be gently inverted 180° and back several times (5-6X for Silica, 8-10X for EDTA)
- Centrifugation conditions: ≤1300 g for 10 minutes at room temperature (25°C)
Specifications
Specifications
| Product Type | Vacutainer Crossmatch Tube |
| Additive | Silica (Clot Activator) |
| Material | PET |
| Sterility | Sterile |
| Closure Type | Hemogard |
| Capacity (Metric) | 6 mL |
| For Use With (Equipment) | Vacutainer™ Needle and Tube Holder |
| Closure Color | Pink |
| Tube Size | 13 x 100 mm |
| For Use With (Application) | Used to obtain either a serum sample or a whole blood or EDTA plasma sample |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction