missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Glass Aprotinine K3EDTA Tube

Product Code. 12997666
Change view
Click to view available options
Capacity (Metric):
5 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12997666 5 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12997666 Supplier BD Supplier No. 361017
  • ONLINE EXCLUSIVE PRICE
    £2635.15
    £1948.66 / Pack of 1000
    Save £686.49
    26% Off
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Available for assays that require a proteolytic enzyme inhibitor. BD Vacutainer™ Glass Aprotinine K3EDTA Tube is a 5mL tube for whole blood or EDTA plasma sample collection. It contains a K3 potassium salt solution and is identified by a pink-peach Hemogard™ closure.

The BD Vacutainer™ system is optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process. Collection tubes have the precise vacuum level to collect the needed sample size, and are designed to work with Vacutainer double-sided needles and tube holders
  • EDTA tubes are used to obtain a whole blood or EDTA plasma sample
  • K3 Potassium Salt of EDTA (Ethylene Diamine Tetra Acetic acid) is used as an anticoagulant
  • K3EDTA is a liquid solution in the BD Vacutainer™ Glass Tubes
  • Aprotinine K3EDTA tube is used for assays that require a proteolytic enzyme inhibitor
  • A pink-peach cap identifies Aprotinine K3EDTA tubes, to differentiate it from regular EDTA tubes
    • The cap color does not follow the color code described in ISO 6710:1995
    • Ensure that users are aware, so the wrong tube is not used for a test, which may result in analytical error
TRUSTED_SUSTAINABILITY

Specifications

Product Type Aprotinine K3 EDTA Tube
Additive K3 EDTA
Material Glass
Sterility Sterile
Closure Type Hemogard
Capacity (Metric) 5 mL
Closure Color Pink-Peach
Tube Size 13 x 75 mm
For Use With (Application) Used to obtain a whole blood or EDTA plasma sample
Label Type Paper
Certifications/Compliance CE labeled
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.