missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Vacuette Tube™, Premium, Red Cap, Black Ring, CAT Serum Clot Activator, Sterile

Product Code. 15402540
Change view
Click to view available options
Capacity (Metric):
3 mL
Closure Color:
Red with Black Ring
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Unit Size
15402540 3 mL Red with Black Ring Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15402540 Supplier Greiner Bio-One Supplier No. 454095
  • ONLINE EXCLUSIVE PRICE
    £216.00
    £198.48 / Pack of 50
    Save £17.52
    8% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Includes inner wall coated with microscopic silica particles to activate coagulation.

  • The inner wall of the serum tube is specially coated with microscopic silica particles, which activate the coagulation process.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Vacuette Tube
Additive CAT Serum Clot Activator
Sterility Sterile
Capacity (Metric) 3 mL
Closure Color Red with Black Ring
Length (Metric) 75 mm
Diameter (Metric) Outer 13 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.