missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Vacuette Low Draw Volume Tube

Product Code. 15474944
Click to view available options
Closure Color:
Lavender
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 15474944

Brand: Greiner Bio-One 454024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Product Type Vacuette Tube
Additive K2 EDTA
Sterility Sterile
Closure Type Cap
Capacity (Metric) 2 mL
Closure Color Lavender
Tube Size 13 x 75 mm
For Use With (Application) For the examin ation of whole blood in haematology
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.