missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Vacuette™ CDPA Blood Collection Tube

Product Code. 10402695
Change view
Click to view available options
Capacity (Metric):
9 mL
Closure Color:
Yellow with Black Ring
Unit Size:
Pack of 1200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color unitSize
10402695 9 mL Yellow with Black Ring Pack of 1200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10402695 Supplier Greiner Bio-One Supplier No. 455056
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Vacuette™ Blood Collection Tubes have set the standard in today‘s marketplace. They provide improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass.

  • Made from virtually unbreakable PET plastic
  • Optically clear as glass
  • Contain CPDA solution (citrate phosphate dextrose adenine)
  • Suitable for blood group typing and cell conservation thanks to the cell-stabilising properties
  • Improved safety and hygiene during specimen collection procedures
TRUSTED_SUSTAINABILITY

Specifications

Additive CPDA Solution (Citrate Phosphate Dextrose Adenine)
Material PET
Sterility Sterile
Closure Type Push Cap
Capacity (Metric) 9 mL
Closure Color Yellow with Black Ring
Tube Size 16 x 100 mm
For Use With (Application) Blood group typing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.