missing translation for 'onlineSavingsMsg'
Learn More

Vactosertib, MedChemExpress

Product Code. 16480428
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480428 5 mg 5mg
16447968 10 mg 10mg
16447756 100 mg 100mg
16443897 50 mg 50mg
16423716 25 mg 25mg
16400336 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16480428 Supplier Medchem Express Supplier No. HY19928
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Vactosertib (EW-7197) is a potent, orally active and ATP-competitive activin receptor-like kinase 5 (ALK5) inhibitor with an IC50 of 12.9 nM. Vactosertib also inhibits ALK2 and ALK4 (IC50 of 17.3 nM) at nanomolar concentrations. Vactosertib has potently antimetastatic activity and anticancer effect.

Vactosertib (EW-7197) is a potent, orally active and ATP-competitive activin receptor-like kinase 5 (ALK5) inhibitor with an IC50 of 12.9 nM. Vactosertib also inhibits ALK2 and ALK4 (IC50 of 17.3 nM) at nanomolar concentrations. Vactosertib has potently antimetastatic activity and anticancer effect.

Specifications

Chemical Name or Material Vactosertib
CAS 1352608-82-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H18FN7
Quantity 5 mg
Synonym EW-7197 TEW-7197
Solubility Information DMSO : 100 mg/mL (250.36 mM; Need ultrasonic)
SMILES CC1=CC=CC(C2=C(C3=CN4C(C=C3)=NC=N4)NC(CNC5=CC=CC=C5F)=N2)=N1
Molecular Weight (g/mol) 399.42
Formula Weight 399.42
Percent Purity 98.27%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.