missing translation for 'onlineSavingsMsg'
Learn More

Vactosertib Hydrochloride, MedChemExpress

Product Code. 16453897
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453897

Brand: Medchem Express HY19928A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vactosertib Hydrochloride (EW-7197 Hydrochloride) is a potent, orally active and ATP-competitive activin receptor-like kinase 5 (ALK5) inhibitor with an IC50 of 12.9 nM. Vactosertib Hydrochloride also inhibits ALK2 and ALK4 (IC50 of 17.3 nM) at nanomolar concentrations. Vactosertib Hydrochloride has potently antimetastatic activity and anticancer effect.

Vactosertib Hydrochloride (EW-7197 Hydrochloride) is a potent, orally active and ATP-competitive activin receptor-like kinase 5 (ALK5) inhibitor with an IC50 of 12.9 nM. Vactosertib Hydrochloride also inhibits ALK2 and ALK4 (IC50 of 17.3 nM) at nanomolar concentrations. Vactosertib Hydrochloride has potently antimetastatic activity and anticancer effect.

Specifications

Chemical Name or Material Vactosertib Hydrochloride
CAS 1352610-25-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H19ClFN7
Quantity 50 mg
Synonym EW-7197 HydrochlorideTEW-7197 Hydrochloride
Solubility Information H2O : 50 mg/mL (114.71 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (114.71 mM; Need ultrasonic)
SMILES CC1=CC=CC(C2=C(C3=CN4C(C=C3)=NC=N4)NC(CNC5=CC=CC=C5F)=N2)=N1.Cl
Molecular Weight (g/mol) 435.88
Formula Weight 435.88
Percent Purity 96.24%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.