missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vactosertib Hydrochloride, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Description
Vactosertib Hydrochloride (EW-7197 Hydrochloride) is a potent, orally active and ATP-competitive activin receptor-like kinase 5 (ALK5) inhibitor with an IC50 of 12.9 nM. Vactosertib Hydrochloride also inhibits ALK2 and ALK4 (IC50 of 17.3 nM) at nanomolar concentrations. Vactosertib Hydrochloride has potently antimetastatic activity and anticancer effect.
Specifications
Specifications
| Chemical Name or Material | Vactosertib Hydrochloride |
| CAS | 1352610-25-3 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C22H19ClFN7 |
| Quantity | 5 mg |
| Synonym | EW-7197 HydrochlorideTEW-7197 Hydrochloride |
| Solubility Information | H2O : 50 mg/mL (114.71 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (114.71 mM; Need ultrasonic) |
| SMILES | CC1=CC=CC(C2=C(C3=CN4C(C=C3)=NC=N4)NC(CNC5=CC=CC=C5F)=N2)=N1.Cl |
| Molecular Weight (g/mol) | 435.88 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction