missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAChT/SLC18A3 Antibody (S6-38), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59378JF549
This item is not returnable.
View return policy
Description
VAChT/SLC18A3 Monoclonal antibody specifically detects VAChT/SLC18A3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| VAChT/SLC18A3 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter | |
| Synthetic peptide amino acids 521-532 of human VAChT | |
| 0.1 mL | |
| ABC Transporters, Neuronal Cell Markers, Neuroscience | |
| 6572 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunofluorescence | |
| S6-38 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction