missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAChT/SLC18A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£175.00 - £439.00
Specifications
| Antigen | VAChT/SLC18A3 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18072674
|
Novus Biologicals
NB100-91348-0.025ML |
0.025 mL |
£175.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18726533
|
Novus Biologicals
NB100-91348 |
£439.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
VAChT/SLC18A3 Polyclonal specifically detects VAChT/SLC18A3 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| VAChT/SLC18A3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter | |
| SLC18A3 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Q16572 | |
| 6572 | |
| IgG | |
| Unpurified | |
| Specific for VACHT. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title