Learn More
Invitrogen™ VAChT Recombinant Rabbit Monoclonal Antibody (HL2900)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556912
Product Code. 30276113
  • £434.00 / 100µL
Description
VAChT Recombinant Monoclonal Antibody for IHC (P)
This gene is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VAChT
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
O35304, Q62666
SLC18A3
Recombinant protein encompassing a sequence within the Intracellular domain of human Vesicular Acetylcholine Transporter. The exact sequence is proprietary.
100 μL
Primary
Mouse, Rat
Antibody
IgG
Immunohistochemistry (Paraffin)
HL2900
Unconjugated
SLC18A3
MGC12716; rVAT; SLC18A3; solute carrier family 18 (vesicular acetylcholine transporter), member 3; solute carrier family 18 (vesicular acetylcholine), member 3; solute carrier family 18 (vesicular monoamine) member 3; solute carrier family 18 (vesicular monoamine), member 3; solute carrier family 18 member 3; solute carrier family 18 member A3; solute carrier family 18, member 3; VAChT; VAT; vesicular acetylcholine transporter
Rabbit
Protein A
RUO
20508, 60422
-20°C, Avoid Freeze/Thaw Cycles