missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAChT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120421
This item is not returnable.
View return policy
Description
Positive test controls include: Mouse brain, Rat brain. The target is usually found in the following locations: Membrane, Multi-pass membrane protein. Immunogen sequence: GLLTRSRSER DVLLDEPPQG LYDAVRLRER PVSGQDGEPR SPPGPFDACE DDYNYYYTRS.
This gene is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene.
Specifications
| VAChT | |
| Polyclonal | |
| Unconjugated | |
| SLC18A3 | |
| MGC12716; rVAT; SLC18A3; solute carrier family 18 (vesicular acetylcholine transporter), member 3; solute carrier family 18 (vesicular acetylcholine), member 3; solute carrier family 18 (vesicular monoamine) member 3; solute carrier family 18 (vesicular monoamine), member 3; solute carrier family 18 member 3; solute carrier family 18 member A3; solute carrier family 18, member 3; VAChT; VAT; vesicular acetylcholine transporter | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20508, 60422, 6572 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 2.51 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O35304, Q16572, Q62666 | |
| SLC18A3 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction